Anti-HTR1A Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB121032-200EA 550 EUR
ABCAAB121032-200
Anti-HTR1A Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HTR1A (5-hydroxytryptamine (serotonin) receptor 1A, G protein-coupled)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR