Anti-PYY Mouse Monoclonal Antibody (Biotin)

Supplier: Abcam
ABCAAB121070-50EA 790 EUR
ABCAAB121070-50
Anti-PYY Mouse Monoclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: PYY (peptide YY)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR