Anti-ZNF589 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB121173-100EA 530 EUR
ABCAAB121173-100
Anti-ZNF589 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZNF589 (zinc finger protein 589)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR