Anti-SNN Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB121330-100EA 520 EUR
ABCAAB121330-100
Anti-SNN Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SNN (stannin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR