Anti-FUT11 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB121411-100EA 520 EUR
ABCAAB121411-100
Anti-FUT11 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FUT11 (fucosyltransferase 11 (alpha (1,3) fucosyltransferase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR