Anti-NUSAP1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB121416-100EA 530 EUR
ABCAAB121416-100
Anti-NUSAP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NUSAP1 (nucleolar and spindle associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR