Anti-STARD3NL Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB121592-100EA 530 EUR
ABCAAB121592-100
Anti-STARD3NL Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: STARD3NL (STARD3 N-terminal like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR