Anti-SIKE1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB121860-100EA 530 EUR
ABCAAB121860-100
Anti-SIKE1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SIKE1 (suppressor of IKBKE 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR