Anti-DNAH8 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB121989-100EA 520 EUR
ABCAAB121989-100
Anti-DNAH8 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DNAH8 (dynein, axonemal, heavy chain 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR