Anti-HSD17B13 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB122036-100EA 510 EUR
ABCAAB122036-100
Anti-HSD17B13 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HSD17B13 (hydroxysteroid (17-beta) dehydrogenase 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR