Anti-ZZEF1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB122194-100EA 520 EUR
ABCAAB122194-100
Anti-ZZEF1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZZEF1 (zinc finger, ZZ-type with EF-hand domain 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR