Anti-DCAF16 Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB122795-100EA 520 EUR
ABCAAB122795-100
Anti-DCAF16 Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: DCAF16 (DDB1 and CUL4 associated factor 16)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR