Anti-ATF6 Mouse monoclonal antibody unconjugated [clone: 1-7]

Supplier: Abcam
ABCAAB122897-50EA 660 EUR
ABCAAB122897-50
Anti-ATF6 Mouse monoclonal antibody unconjugated [clone: 1-7]
Antibodies
Type: Primary
Antigen: ATF6 (activating transcription factor 6)
Clonality: monoclonal
Clone: 42011
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR