Anti-DGAT1 Goat polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB122924-50EA 0 EUR
ABCAAB122924-50
Anti-DGAT1 Goat polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: DGAT1 (diacylglycerol O-acyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR