Anti-IgG Mouse Monoclonal Antibody [clone: R10Z8E9]
Supplier: Abcam
ABCAAB124055-100EA 530 EUR
ABCAAB124055-100
Anti-IgG Mouse Monoclonal Antibody [clone: R10Z8E9]
Antibodies
Type: Primary
Antigen: IgG (IgG)
Clonality: monoclonal
Clone: R10Z8E9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human