Anti-THAP11 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB124204-100EA 495 EUR
ABCAAB124204-100
Anti-THAP11 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: THAP11 (THAP domain containing 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR