Anti-Fast Myosin Skeletal Heavy chain Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB124205-100EA 540 EUR
ABCAAB124205-100
Anti-Fast Myosin Skeletal Heavy chain Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: Fast Myosin Skeletal Heavy chain
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR