Anti-HSPA9 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB124382-50EA 560 EUR
ABCAAB124382-50
Anti-HSPA9 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HSPA9 (heat shock 70kDa protein 9 (mortalin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR