Anti-DDB1 Rabbit monoclonal antibody unconjugated [clone: EPR6088]

Supplier: Abcam
ABCAAB124672-100EA 560 EUR
ABCAAB124672-100
Anti-DDB1 Rabbit monoclonal antibody unconjugated [clone: EPR6088]
Antibodies
Type: Primary
Antigen: DDB1 (damage-specific DNA binding protein 1, 127kDa)
Clonality: monoclonal
Clone: EPR6088
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR