Anti-RETN Rabbit Monoclonal Antibody [clone: EP4738]

Supplier: Abcam
ABCAAB124681-100EA 525 EUR
ABCAAB124681-100
Anti-RETN Rabbit Monoclonal Antibody [clone: EP4738]
Antibodies
Type: Primary
Antigen: RETN (resistin)
Clonality: monoclonal
Clone: EP4738
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR