Anti-TAB3 Rabbit Monoclonal Antibody [clone: EPR5965]

Supplier: Abcam
ABCAAB124723-100EA 505 EUR
ABCAAB124723-100
Anti-TAB3 Rabbit Monoclonal Antibody [clone: EPR5965]
Antibodies
Type: Primary
Antigen: TAB3 (TGF-beta activated kinase 1/MAP3K7 binding protein 3)
Clonality: monoclonal
Clone: EPR5965
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR