Anti-TRIM27 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB124754-100EA 560 EUR
ABCAAB124754-100
Anti-TRIM27 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TRIM27 (tripartite motif containing 27)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR