Anti-RAD52 Rabbit Monoclonal Antibody [clone: EPR3464]

Supplier: Abcam
ABCAAB124971-100EA 480 EUR
ABCAAB124971-100
Anti-RAD52 Rabbit Monoclonal Antibody [clone: EPR3464]
Antibodies
Type: Primary
Antigen: RAD52 (RAD52 homolog (S. cerevisiae))
Clonality: monoclonal
Clone: EPR3464
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR