Anti-Retinoid X Receptor alpha Rabbit Monoclonal Antibody [clone: EPR7106]

Supplier: Abcam
ABCAAB125001-100EA 560 EUR
ABCAAB125001-100
Anti-Retinoid X Receptor alpha Rabbit Monoclonal Antibody [clone: EPR7106]
Antibodies
Type: Primary
Antigen: Retinoid X Receptor alpha
Clonality: monoclonal
Clone: EPR7106
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR