Anti-Lipoprotein a Rabbit Monoclonal Antibody [clone: EPR6474]

Supplier: Abcam
ABCAAB125014-100EA 500 EUR
ABCAAB125014-100
Anti-Lipoprotein a Rabbit Monoclonal Antibody [clone: EPR6474]
Antibodies
Type: Primary
Antigen: Lipoprotein a
Clonality: monoclonal
Clone: EPR6474
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR