Anti-AQP4 Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB125049-100EA 550 EUR
ABCAAB125049-100
Anti-AQP4 Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: AQP4 (aquaporin 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR