Anti-CTSB Rabbit Monoclonal Antibody [clone: EPR4323]

Supplier: Abcam
ABCAAB125067-100EA 560 EUR
ABCAAB125067-100
Anti-CTSB Rabbit Monoclonal Antibody [clone: EPR4323]
Antibodies
Type: Primary
Antigen: CTSB (cathepsin B)
Clonality: monoclonal
Clone: EPR4323
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR