Anti-VIPAS39 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB125084-100EA 510 EUR
ABCAAB125084-100
Anti-VIPAS39 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: VIPAS39 (VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR