Anti-CDC6 Rabbit Polyclonal Antibody
Supplier: Abcam
ABCAAB125195-100EA 550 EUR
ABCAAB125195-100
Anti-CDC6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDC6 (cell division cycle 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse