Anti-HSPA8 Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB125307-100EA 545 EUR
ABCAAB125307-100
Anti-HSPA8 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: HSPA8 (heat shock 70kDa protein 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR