Anti-TP53INP1 Rabbit Polyclonal Antibody

ABCAAB125960-100EA 395 EUR
ABCAAB125960-100
Anti-TP53INP1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: TP53INP1 (tumor protein p53 inducible nuclear protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR