Anti-KBTBD4 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB126078-50EA 510 EUR
ABCAAB126078-50
Anti-KBTBD4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KBTBD4 (kelch repeat and BTB (POZ) domain containing 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR