Anti-SH3BP5 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB126231-50EA 530 EUR
ABCAAB126231-50
Anti-SH3BP5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SH3BP5 (SH3-domain binding protein 5 (BTK-associated))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR