Anti-DCDC2C Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB126564-100EA 520 EUR
ABCAAB126564-100
Anti-DCDC2C Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: DCDC2C (doublecortin domain containing 2C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR