Anti-SUZ12 Mouse Monoclonal Antibody [clone: SUZ220A]

Supplier: Abcam
ABCAAB126577-100EA 495 EUR
ABCAAB126577-100
Anti-SUZ12 Mouse Monoclonal Antibody [clone: SUZ220A]
Antibodies
Type: Primary
Antigen: SUZ12 (SUZ12 polycomb repressive complex 2 subunit)
Clonality: monoclonal
Clone: SUZ220A
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR