Anti-ALKBH1 Rabbit Monoclonal Antibody [clone: EPR6176]

Supplier: Abcam
ABCAAB126596-100EA 580 EUR
ABCAAB126596-100
Anti-ALKBH1 Rabbit Monoclonal Antibody [clone: EPR6176]
Antibodies
Type: Primary
Antigen: ALKBH1 (alkB, alkylation repair homolog 1 (E. coli))
Clonality: monoclonal
Clone: EPR6176
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR