Anti-HLA F Rabbit Monoclonal Antibody [clone: EPR6803]

Supplier: Abcam
ABCAAB126624-100EA 505 EUR
ABCAAB126624-100
Anti-HLA F Rabbit Monoclonal Antibody [clone: EPR6803]
Antibodies
Type: Primary
Antigen: HLA F
Clonality: monoclonal
Clone: EPR6803
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR