Anti-IFIH1 Rabbit Monoclonal Antibody [clone: EPR6743]

Supplier: Abcam
ABCAAB126630-100EA 605 EUR
ABCAAB126630-100
Anti-IFIH1 Rabbit Monoclonal Antibody [clone: EPR6743]
Antibodies
Type: Primary
Antigen: IFIH1 (interferon induced with helicase C domain 1)
Clonality: monoclonal
Clone: EPR6743
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR