Anti-Prostaglandin F2 alpha Receptor Rabbit Monoclonal Antibody [clone: EPR5844]

Supplier: Abcam
ABCAAB126709-100EA 525 EUR
ABCAAB126709-100
Anti-Prostaglandin F2 alpha Receptor Rabbit Monoclonal Antibody [clone: EPR5844]
Antibodies
Type: Primary
Antigen: Prostaglandin F2 alpha Receptor
Clonality: monoclonal
Clone: EPR5844
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR