Anti-CDC45 Rabbit Monoclonal Antibody [clone: EPR5759]
Supplier: Abcam
ABCAAB126762-100EA 485 EUR
ABCAAB126762-100
Anti-CDC45 Rabbit Monoclonal Antibody [clone: EPR5759]
Antibodies
Type: Primary
Antigen: CDC45 (cell division cycle 45)
Clonality: monoclonal
Clone: EPR5759
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now