Anti-CTCFL Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB126778-100EA 530 EUR
ABCAAB126778-100
Anti-CTCFL Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: CTCFL (CCCTC-binding factor (zinc finger protein)-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR