Anti-FMO3 Rabbit monoclonal antibody unconjugated [clone: EPR6967]

Supplier: Abcam
ABCAAB126790-100EA 535 EUR
ABCAAB126790-100
Anti-FMO3 Rabbit monoclonal antibody unconjugated [clone: EPR6967]
Antibodies
Type: Primary
Antigen: FMO3 (flavin containing monooxygenase 3)
Clonality: monoclonal
Clone: EPR6967
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR