Anti-ALP Mouse Monoclonal Antibody [clone: 2F4]

Supplier: Abcam
ABCAAB126820-100EA 495 EUR
ABCAAB126820-100
Anti-ALP Mouse Monoclonal Antibody [clone: 2F4]
Antibodies
Type: Primary
Antigen: ALP (Alkaline Phosphatase)
Clonality: monoclonal
Clone: 2F4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR