Anti-DEFB119 Goat polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB126867-200EA 350 EUR
ABCAAB126867-200
Anti-DEFB119 Goat polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: DEFB119 (defensin, beta 119)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR