Anti-CYP1A1 Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB126887-200EA 550 EUR
ABCAAB126887-200
Anti-CYP1A1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CYP1A1 (cytochrome P450, family 1, subfamily A, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR