Anti-RNF40 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB126959-50EA 560 EUR
ABCAAB126959-50
Anti-RNF40 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RNF40 (ring finger protein 40, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR