Anti-TNPO2 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB127165-50EA 530 EUR
ABCAAB127165-50
Anti-TNPO2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TNPO2 (transportin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR