Anti-CCDC50 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB127169-50EA 510 EUR
ABCAAB127169-50
Anti-CCDC50 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CCDC50 (coiled-coil domain containing 50)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR