Anti-GNPNAT1 Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB127697-50EA 510 EUR
ABCAAB127697-50
Anti-GNPNAT1 Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: GNPNAT1 (glucosamine-phosphate N-acetyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR