Anti-PTTG1IP Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB128040-50EA 550 EUR
ABCAAB128040-50
Anti-PTTG1IP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PTTG1IP (pituitary tumor-transforming 1 interacting protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR